Learn More
Sino Biological Protein LASV-NP Biotin 20UG
Catalog No. 5001074121
Encompass_Preferred
A DNA sequence encoding the Lassa virus (strain Mouse/Sierra Leone/Josiah/1976) NP (P13699) (Met1-Leu569) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-74121 Supplier Sino Biological Supplier No. 41179V49EB20
A DNA sequence encoding the Lassa virus (strain Mouse/Sierra Leone/Josiah/1976) NP (P13699) (Met1-Leu569) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed