Learn More
Sino Biological Recombinant Legionella pneumophila LegK4 Protein (Kinase Domain, GST-TEV Tag), Active 10 µg
Catalog No. 5000656701
Encompass_Preferred
Recombinant Legionella pneumophila (strain Lens) LegK4 (73-394; kinase domain) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST-TEV tag. This gene accession number is Q5WZW9.
Catalog No. 50-006-56701 Supplier Sino Biological Supplier No. L4111G10
Recombinant Legionella pneumophila (strain Lens) LegK4 (73-394; kinase domain) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST-TEV tag. This gene accession number is Q5WZW9.