Learn More
Showa Denko America Inc PROTEIN LW-803
Supplier:  Showa Denko America Inc F6989303
Encompass_Preferred
Pore size specifically controlled for analyzing proteins with molecular weights of several hundred thousand Daltons; optimized for light scattering detection, high performance analysis of antibody drugs and various proteins, high lot-to-lot reproducibility, fulfills USP L20, L33, and L59 requirements, MW range = 5,000 to 100,000 Daltons, exclusion limit = 170,000 Daltons, dimensions 8.0 mm ID x 300 mm L
Catalog No. 50-161-3281