Learn More
Biotang Inc Prestained Protein Marker (10-240 Kda),Three color bands(Green, Blue, Red). Ready-to-Use no need to boil.Broad range size estimation(11-240kDa). Highly purified and blended 11 recombinant proteins 2x250ul. Store at 2-8 deg.C
Catalog No. 507514908 Shop All BIOTANG Inc Products
Encompass_Preferred
Prestained Protein Marker (10-240 Kda),Three color bands(Green, Blue, Red). Ready-to-Use no need to boil.Broad range size estimation(11-240kDa). Highly purified and blended 11 recombinant proteins 2x250ul. Store at 2-8 deg.C
Catalog No. 50-751-4908 Supplier Biotang Inc Supplier No. P8502050
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Prestained Protein Marker (10-240 Kda),Three color bands(Green, Blue, Red). Ready-to-Use no need to boil.Broad range size estimation(11-240kDa). Highly purified and blended 11 recombinant proteins 2x250ul. Store at 2-8 deg.C