Learn More
ACROBiosystems Star Ribbon Pre-stained Protein Marker (8-180 kDa)
Catalog No. 502331361
Encompass_Preferred
This product is a mixture of ten highly purified pre-stained proteins ranging from 8 kDa to 180 kDa. The 72 kDa band is orange, 8 kDa band is green, and other bands are blue. It is designed for observing protein separation during SDS-PAGE, verifying western transfer efficiency on membranes, and approximating the size of proteins.
Catalog No. 50-233-1361 Supplier ACROBiosystems Supplier No. PPMM10012500UL
This product is a mixture of ten highly purified pre-stained proteins ranging from 8 kDa to 180 kDa. The 72 kDa band is orange, 8 kDa band is green, and other bands are blue. It is designed for observing protein separation during SDS-PAGE, verifying western transfer efficiency on membranes, and approximating the size of proteins.