Learn More
Rockland Immunochemicals Protein Molecular Weight Marker (14 - 120kDa) - MB-201-0200
SDP
Catalog No. 501059076
Encompass_Preferred
This protein molecular weight marker is a cocktail containing seven purified proteins that range in size from 14 to 120 kDa and thus span a molecular weight range common to most proteins and protein subunits.  The protein mixture has been formulated to yield well-defined bands after SDS–polyacrylamide gel electrophoresis and staining with Coomassie blue or other common staining techniques.
Catalog No. 50-105-9076 Supplier Rockland Immunochemicals Supplier No. MB2010200
This protein molecular weight marker is a cocktail containing seven purified proteins that range in size from 14 to 120 kDa and thus span a molecular weight range common to most proteins and protein subunits.  The protein mixture has been formulated to yield well-defined bands after SDS–polyacrylamide gel electrophoresis and staining with Coomassie blue or other common staining techniques.