Learn More
Sino Biological Protein Mouse BMP4 ET-FREE 1MG
Catalog No. 5001074155
Encompass_Preferred
A DNA sequence encoding the mouse BMP4 (NP 031580 2) (Ser293-Arg408) was expressed with the Fc region of rabbit IgG at the N-terminus
Catalog No. 50-010-74155 Supplier Sino Biological Supplier No. 50439M15HUE1
A DNA sequence encoding the mouse BMP4 (NP 031580 2) (Ser293-Arg408) was expressed with the Fc region of rabbit IgG at the N-terminus