Learn More
Sino Biological Protein Mouse CD8A ET-FREE 1MG
Catalog No. 5001073792
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD8a (NP 001074579 1) (Met 1-Tyr 196) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73792 Supplier Sino Biological Supplier No. 50389M08HUE1
A DNA sequence encoding the extracellular domain of mouse CD8a (NP 001074579 1) (Met 1-Tyr 196) was expressed with a C-terminal polyhistidine tag