Learn More
Sino Biological Protein Mouse CES2 1MG
Catalog No. 5001074158
Encompass_Preferred
A DNA sequence encoding the Mouse carboxylesterase 2/CES2 (NP 663578 1) (Met1-Lys557) was expressed fused with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74158 Supplier Sino Biological Supplier No. 50515M08H11
A DNA sequence encoding the Mouse carboxylesterase 2/CES2 (NP 663578 1) (Met1-Lys557) was expressed fused with a polyhistidine tag at the C-terminus