Learn More
Sino Biological Protein Mouse CTLA4 Biotin 20U
Catalog No. 5001074157
Encompass_Preferred
A DNA sequence encoding the Mouse CTLA-4 (NP 033973 2) (Glu36-Asp161) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-74157 Supplier Sino Biological Supplier No. 50503M49H1B20
A DNA sequence encoding the Mouse CTLA-4 (NP 033973 2) (Glu36-Asp161) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed