Learn More
Sino Biological Protein Mouse DSC1 100UG
Catalog No. 5001073960
Encompass_Preferred
A DNA sequence encoding the Mouse DSC1 (P55849) (Arg135-Lys691) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-73960 Supplier Sino Biological Supplier No. 5A0409M08H100
A DNA sequence encoding the Mouse DSC1 (P55849) (Arg135-Lys691) was expressed with a polyhistidine tag at the C-terminus