Learn More
Sino Biological Recombinant Mouse Tissue Factor Protein (His Tag), HPLC-verified, Endotoxin-Free 100 µg
Catalog No. 5000656605
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse F3 (NP_034301.3) (Met 1-Glu 251) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-006-56605 Supplier Sino Biological Supplier No. 50413M08HUE100
A DNA sequence encoding the extracellular domain of mouse F3 (NP_034301.3) (Met 1-Glu 251) was expressed, with a C-terminal polyhistidine tag.