Learn More
Sino Biological Protein Mouse FGF18 100UG
Catalog No. 5000699179
Encompass_Preferred
A DNA sequence encoding the mouse FGF18 (NP_032031.1) (Met 1-Gly 207) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-99179 Supplier Sino Biological Supplier No. 50177M08HUE100
A DNA sequence encoding the mouse FGF18 (NP_032031.1) (Met 1-Gly 207) was fused with a polyhistidine tag at the C-terminus.