Learn More
Sino Biological Recombinant Mouse IGSF11 Protein (hFc Tag), HPLC-verified 100 µg
Catalog No. 5000656587
Encompass_Preferred
A DNA sequence encoding the Mouse IGSF11 (P0C673) (Met1-Val240) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-006-56587 Supplier Sino Biological Supplier No. 5A6052M02H100
A DNA sequence encoding the Mouse IGSF11 (P0C673) (Met1-Val240) was expressed, fused with the Fc region of human IgG1 at the C-terminus.