Learn More
Sino Biological Protein Mouse IL25 ET-FREE 1MG
Catalog No. 5001073667
Encompass_Preferred
A DNA sequence encoding the mature form of mouse IL25 (NP 542767 1) (Val 17-Ala 169) was expressed with a N-terminal polyhistidine tag
Catalog No. 50-010-73667 Supplier Sino Biological Supplier No. 50138M07HUE1
A DNA sequence encoding the mature form of mouse IL25 (NP 542767 1) (Val 17-Ala 169) was expressed with a N-terminal polyhistidine tag