Learn More
Sino Biological Protein Mouse IL31 ET-FREE 1MG
Catalog No. 5001074105
Encompass_Preferred
A DNA sequence encoding the mouse Il31 (NP 083870 1) (Met1-Cys163) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74105 Supplier Sino Biological Supplier No. 51070M08HUE1
A DNA sequence encoding the mouse Il31 (NP 083870 1) (Met1-Cys163) was expressed with a polyhistidine tag at the C-terminus