Learn More
Sino Biological Recombinant Mouse Integrin alpha 3 beta 1 Protein (Flag & His Tag), HPLC-verified 100 µg
Catalog No. 5000656847
Encompass_Preferred
A DNA sequence encoding the Mouse ITGA3 Q62470)(Phe33-Glu993) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the Mouse ITGB1 (P09055) (Gln21-Asp728) was fused with a flag tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the Mouse Integrin alpha 3 beta 1 Heterodimer was purified.
Catalog No. 50-006-56847 Supplier Sino Biological Supplier No. CT180M0825H100
A DNA sequence encoding the Mouse ITGA3 Q62470)(Phe33-Glu993) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the Mouse ITGB1 (P09055) (Gln21-Asp728) was fused with a flag tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the Mouse Integrin alpha 3 beta 1 Heterodimer was purified.