Learn More
Sino Biological Protein Mouse ITPRIPL1 100UG
Catalog No. 5000698854
Encompass_Preferred
A DNA sequence encoding the Mouse ITPRIPL1 (A2ASA8) (Asp23-Gly96) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-98854 Supplier Sino Biological Supplier No. 5A5418M08HUE100
A DNA sequence encoding the Mouse ITPRIPL1 (A2ASA8) (Asp23-Gly96) was expressed with a polyhistidine tag at the C-terminus.