Learn More
Sino Biological Protein Mouse ITPRIPL1 1MG
Catalog No. 5000698689
Encompass_Preferred
A DNA sequence encoding the Mouse ITPRIPL1 (A2ASA8) (Asp23-Gly96) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-98689 Supplier Sino Biological Supplier No. 5A5418M08HUE1
A DNA sequence encoding the Mouse ITPRIPL1 (A2ASA8) (Asp23-Gly96) was expressed with a polyhistidine tag at the C-terminus.