Learn More
Sino Biological Protein Mouse KLB 100UG
Catalog No. 5000698678
Encompass_Preferred
A DNA sequence encoding the Mouse KLB (NP_112457.1) (Phe53-Leu995) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-98678 Supplier Sino Biological Supplier No. 50661M08H1UE100
A DNA sequence encoding the Mouse KLB (NP_112457.1) (Phe53-Leu995) was expressed with a polyhistidine tag at the C-terminus.