Learn More
Sino Biological Protein Mouse NOG ET-FREE 1MG
Catalog No. 5001073796
Encompass_Preferred
A DNA sequence encoding the mouse NOG (P97466) (Met1-Cys232) was fused with the Fc region of human IgG1 at the C-terminus
Catalog No. 50-010-73796 Supplier Sino Biological Supplier No. 50688M02HUE1
A DNA sequence encoding the mouse NOG (P97466) (Met1-Cys232) was fused with the Fc region of human IgG1 at the C-terminus