Learn More
Sino Biological Recombinant Mouse NOTCH1 Protein (His Tag), HPLC-verified 100 µg
Catalog No. 5000656660
Encompass_Preferred
A DNA sequence encoding the Mouse NOTCH1 (NP_032740.3) (Ala19-Gln526) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-56660 Supplier Sino Biological Supplier No. 50654M08H100
A DNA sequence encoding the Mouse NOTCH1 (NP_032740.3) (Ala19-Gln526) was expressed with a polyhistidine tag at the C-terminus.