Learn More
Sino Biological Recombinant Mouse Nephrin Protein (ECD, His Tag) 1 mg
Catalog No. 5000656676
Encompass_Preferred
A DNA sequence encoding the Mouse Nephrin (Q9QZS7) (Ala36-Leu1052) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-56676 Supplier Sino Biological Supplier No. 57864M08H1
A DNA sequence encoding the Mouse Nephrin (Q9QZS7) (Ala36-Leu1052) was fused with a polyhistidine tag at the C-terminus.