Learn More
Sino Biological Recombinant Mouse Cyclophilin A Protein 1000 µg
Catalog No. 5000656846
Encompass_Preferred
Recombinant full-length mouse Cyclophilin A protein was expressed in E.coli cells using an N-terminal His-bdSUMO tag, and untagged prior to final purification.
Catalog No. 50-006-56846 Supplier Sino Biological Supplier No. C70130U1000
Recombinant full-length mouse Cyclophilin A protein was expressed in E.coli cells using an N-terminal His-bdSUMO tag, and untagged prior to final purification.