Learn More
Sino Biological Recombinant Mouse Cyclophilin A Protein 100 µg
Catalog No. 5000656682
Encompass_Preferred
Recombinant full-length mouse Cyclophilin A protein was expressed in E.coli cells using an N-terminal His-bdSUMO tag, and untagged prior to final purification.
Catalog No. 50-006-56682 Supplier Sino Biological Supplier No. C70130U100
Recombinant full-length mouse Cyclophilin A protein was expressed in E.coli cells using an N-terminal His-bdSUMO tag, and untagged prior to final purification.