Learn More
Sino Biological Recombinant Mouse S100A4 Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified 100 µg
Catalog No. 5000656584
Encompass_Preferred
A DNA sequence encoding the Mouse S100A4 (NP_035441.1) (Met1-Lys101) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-56584 Supplier Sino Biological Supplier No. 50484M56EB100
A DNA sequence encoding the Mouse S100A4 (NP_035441.1) (Met1-Lys101) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.