Learn More
Sino Biological Recombinant Mouse SOST/Sclerostin Protein (hFc Tag), HPLC-verified 100 µg
Catalog No. 5000656666
Encompass_Preferred
A DNA sequence encoding the Mouse SOST/Sclerostin (Q99P68) (Gln24-Tyr211) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-006-56666 Supplier Sino Biological Supplier No. 57963M01H100
A DNA sequence encoding the Mouse SOST/Sclerostin (Q99P68) (Gln24-Tyr211) was expressed with the fused Fc region of human IgG1 at the N-terminus.