Learn More
Sino Biological Recombinant Mouse Transferrin Receptor/TFR1/CD71 Protein (His Tag), HPLC-verified 1 mg
Catalog No. 5000656548
Encompass_Preferred
A DNA sequence encoding the Mouse Transferrin Receptor/TFR1/CD71 (Q62351) (Cys89-Phe763) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-006-56548 Supplier Sino Biological Supplier No. 50741M07H21
A DNA sequence encoding the Mouse Transferrin Receptor/TFR1/CD71 (Q62351) (Cys89-Phe763) was expressed with a polyhistidine tag at the N-terminus.