Learn More
Sino Biological Protein Mouse TGFB1 1MG
Catalog No. 5000698845
Encompass_Preferred
A DNA sequence encoding the mouse Latent TGFB1 (P04202) (Met1-Ser390) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-006-98845 Supplier Sino Biological Supplier No. 50698M08HUE1
A DNA sequence encoding the mouse Latent TGFB1 (P04202) (Met1-Ser390) was expressed with a C-terminal polyhistidine tag.