Learn More
Sino Biological Protein Mouse TNC 1MG
Catalog No. 5001073958
Encompass_Preferred
A DNA sequence encoding the Mouse TNC (Q80YX1) (Pro626-Leu1077) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-73958 Supplier Sino Biological Supplier No. 57097M08H1
A DNA sequence encoding the Mouse TNC (Q80YX1) (Pro626-Leu1077) was expressed with a polyhistidine tag at the C-terminus