Learn More
Sino Biological Protein NCAM1 ET-FREE 1MG
Catalog No. 5001073831
Encompass_Preferred
A DNA sequence encoding the human NCAM1 (NP 001070150 1) (Met1-Pro603) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-73831 Supplier Sino Biological Supplier No. 10673H08HUE1
A DNA sequence encoding the human NCAM1 (NP 001070150 1) (Met1-Pro603) was expressed with a polyhistidine tag at the C-terminus