Learn More
Sino Biological Protein NECTIN4 AF488 25UG
Catalog No. 5001073745
Encompass_Preferred
A DNA sequence encoding the human Nectin-4 (NP 112178 2) (Met 1-Ser 349) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 488 (Excitation Max 495 nm Emission Max 519 nm)
Catalog No. 50-010-73745 Supplier Sino Biological Supplier No. 19771H86HSD25
A DNA sequence encoding the human Nectin-4 (NP 112178 2) (Met 1-Ser 349) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 488 (Excitation Max 495 nm Emission Max 519 nm)