Learn More
Sino Biological Protein NECTIN4 AF647 25UG
Catalog No. 5001074031
Encompass_Preferred
A DNA sequence encoding the human Nectin-4 (NP 112178 2) (Met 1-Ser 349) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 647 (Excitation Max 655 nm Emission Max 680 nm)
Catalog No. 50-010-74031 Supplier Sino Biological Supplier No. 19771H86HSG25
A DNA sequence encoding the human Nectin-4 (NP 112178 2) (Met 1-Ser 349) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 647 (Excitation Max 655 nm Emission Max 680 nm)