Learn More
Vector Laboratories Protein-Oligonucleotide Conjugation Kit, 1 Kit
SDP
Catalog No. 502045759
Encompass
S-HyNic (2 X 1 mg)S-4FB (2 x 1mg)Kit Components: 10X Modification Buffer (1.5 mL); 10X Conjugation Buffer (1.5 mL); 10X TurboLINK™ Catalyst Buffer (1.5 mL); 7kDa MWCO, 0.5 mL Thermo Scientific™ Zeba™ Column (4); Anhydrous DMF (1.5 mL); 2-Hydrazinopyridine (2-HP) (0.5 mL); 2-Sulfobenzaldehyde (2-SB) (0.5 mL); 7kDa MWCO 2 mL Zeba Column (2); 10X PBS (1.5 mL); 2 mL Collection Tube (12); Oligo Resuspension Solution (1.0 mL);
Catalog No. 50-204-5759 Supplier Vector Laboratories Supplier No. S90111
S-HyNic (2 X 1 mg)S-4FB (2 x 1mg)Kit Components: 10X Modification Buffer (1.5 mL); 10X Conjugation Buffer (1.5 mL); 10X TurboLINK™ Catalyst Buffer (1.5 mL); 7kDa MWCO, 0.5 mL Thermo Scientific™ Zeba™ Column (4); Anhydrous DMF (1.5 mL); 2-Hydrazinopyridine (2-HP) (0.5 mL); 2-Sulfobenzaldehyde (2-SB) (0.5 mL); 7kDa MWCO 2 mL Zeba Column (2); 10X PBS (1.5 mL); 2 mL Collection Tube (12); Oligo Resuspension Solution (1.0 mL);