Learn More
Waters Corp Protein-Pak Column, 10 μm, 7.8 mm, 300 mm
Catalog No. 50785917 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Protein-Pak 125 Columns support legacy methods, utilizing 10 μm diol-bonded gel filtration materials. These columns effectively resolve biopolymers, including proteins and enzymes, in HPLC applications, distinguishing proteins that differ by as little as 15% in molecular weight.
  • Made with 10 μm diol-bonded gel filtration materials.
  • Resolves proteins that differ in molecular weight by a factor of two.
  • Distinguishes proteins by as little as 15% in molecular weight.
  • Enables separation of biopolymers in HPLC applications.
  • Thoroughly tested in a quality-controlled laboratory.
  • Compatible with water-miscible solvents like methanol and acetonitrile.
  • Stable in normal salt buffer solutions (e.g., sodium sulfate, ammonium acetate, phosphate buffers).
Catalog No. 50-785-917 Supplier Waters Corp Supplier No. WAT084601
Protein-Pak 125 Columns support legacy methods, utilizing 10 μm diol-bonded gel filtration materials. These columns effectively resolve biopolymers, including proteins and enzymes, in HPLC applications, distinguishing proteins that differ by as little as 15% in molecular weight.
  • Made with 10 μm diol-bonded gel filtration materials.
  • Resolves proteins that differ in molecular weight by a factor of two.
  • Distinguishes proteins by as little as 15% in molecular weight.
  • Enables separation of biopolymers in HPLC applications.
  • Thoroughly tested in a quality-controlled laboratory.
  • Compatible with water-miscible solvents like methanol and acetonitrile.
  • Stable in normal salt buffer solutions (e.g., sodium sulfate, ammonium acetate, phosphate buffers).