Learn More
Waters Corp Waters Sentry Guard Cartridge, Diol (OH), 125 Å, 10 μm, 3.9 mm x 20 mm
Catalog No. 50780518 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sentry Guard Cartridge is a protective piece of lab equipment, placed between the injector and the analytical column, to improve efficiency by obstructing chemical impurities and suspended particles. It consists of 10 μm diol-bonded gel filtration materials, protecting 3.9 mm and 4.6 mm I.D. analytical columns. Manufactured in a cGMP, ISO 9001-certified facility, it ensures quality and durability.
  • Protects and preserves analytical columns.
  • Obstructs chemical impurities and suspended particles.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes.
  • Each piece of equipment is tested before approval for use.
  • Increases selectivity, sensitivity, and speed of studies.
Catalog No. 50-780-518 Supplier Waters Corp Supplier No. 186000926
This Sentry Guard Cartridge is a protective piece of lab equipment, placed between the injector and the analytical column, to improve efficiency by obstructing chemical impurities and suspended particles. It consists of 10 μm diol-bonded gel filtration materials, protecting 3.9 mm and 4.6 mm I.D. analytical columns. Manufactured in a cGMP, ISO 9001-certified facility, it ensures quality and durability.
  • Protects and preserves analytical columns.
  • Obstructs chemical impurities and suspended particles.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes.
  • Each piece of equipment is tested before approval for use.
  • Increases selectivity, sensitivity, and speed of studies.