Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Protein-Pak SEC Column, 300 Å, 10 μm

Catalog No. 50785897 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Protein-Pak SEC Column primarily supports legacy methods, utilizing 10 μm diol-bonded gel filtration materials. It is suitable for larger biologically active molecules, resolving proteins that differ by a factor of two or as little as 15% in molecular weight.

  • Composed of 10 μm diol-bonded gel filtration materials.
  • Designed for molecular weight ranges: 10,000 to 400,000 (Native Globular) and 2,000 to 150,000 (Random Coil).
  • Resolves proteins differing by factor of two or as little as 15% in molecular weight.
  • Stable in common salt buffer solutions (e.g., sodium sulfate, phosphate buffers).
  • 0.1 M ionic strength minimizes protein-stationary phase interaction.
  • Shipped in 0.05% sodium azide in a water solution.

Catalog No. 50-785-897 Supplier Waters Corp Supplier No. WAT080013
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Protein-Pak SEC Column primarily supports legacy methods, utilizing 10 μm diol-bonded gel filtration materials. It is suitable for larger biologically active molecules, resolving proteins that differ by a factor of two or as little as 15% in molecular weight.

  • Composed of 10 μm diol-bonded gel filtration materials.
  • Designed for molecular weight ranges: 10,000 to 400,000 (Native Globular) and 2,000 to 150,000 (Random Coil).
  • Resolves proteins differing by factor of two or as little as 15% in molecular weight.
  • Stable in common salt buffer solutions (e.g., sodium sulfate, phosphate buffers).
  • 0.1 M ionic strength minimizes protein-stationary phase interaction.
  • Shipped in 0.05% sodium azide in a water solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.