Learn More
Waters Corp Waters SEC Column, 10 μm, 19 mm, 300 mm, Diol
Catalog No. 50785216 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Protein-Pak Size-Exclusion Columns (SEC) resolve proteins by molecular weight, distinguishing proteins that differ by as little as 15%. Ideal for biopolymers like peptides, polypeptides, steroids, and small proteins, they provide excellent separation for Native Globular 1,000–20,000 and Random Coil 600–8,000 molecular weight ranges.
  • Supports legacy methods with 10 μm diol-bonded gel filtration materials.
  • Available in various pore sizes and configurations.
  • Resolution influenced by sample volume and mass; minimizes sample-stationary phase interaction.
  • Shipped in 100% methanol.
  • Ensures high quality outcomes for liquid chromatography.
  • Produced in specialized facilities adhering to cGMP, ISO 9001, and ISO 3485 standards.
  • Rigorous quality control ensures consistent batch-to-batch performance.
Catalog No. 50-785-216 Supplier Waters Corp Supplier No. WAT025830
Protein-Pak Size-Exclusion Columns (SEC) resolve proteins by molecular weight, distinguishing proteins that differ by as little as 15%. Ideal for biopolymers like peptides, polypeptides, steroids, and small proteins, they provide excellent separation for Native Globular 1,000–20,000 and Random Coil 600–8,000 molecular weight ranges.
  • Supports legacy methods with 10 μm diol-bonded gel filtration materials.
  • Available in various pore sizes and configurations.
  • Resolution influenced by sample volume and mass; minimizes sample-stationary phase interaction.
  • Shipped in 100% methanol.
  • Ensures high quality outcomes for liquid chromatography.
  • Produced in specialized facilities adhering to cGMP, ISO 9001, and ISO 3485 standards.
  • Rigorous quality control ensures consistent batch-to-batch performance.