Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Glass Column, 10 μm, 8 mm × 75 mm

Catalog No. 50785048 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Protein-Pak DEAE Glass Column is designed for protein applications and utilizes 10 μm diol-bonded gel filtration materials.

  • Chemistry: Anion-exchange
  • Separation mode: Ion-exchange
  • Particle substrate: Polymethacrylate
  • pH range: 2 pH to 12 pH
  • Silanol activity: Low
  • Molecular weight range: 1000 to 500000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 1000 Å
  • Format: AP glass column
  • System: HPLC
  • USP classification: L23
  • Inner diameter: 8 mm
  • Length: 75 mm
  • Application: Protein
  • Units per package: 1 pk

Catalog No. 50-785-048 Supplier Waters Corp Supplier No. WAT011783
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Protein-Pak DEAE Glass Column is designed for protein applications and utilizes 10 μm diol-bonded gel filtration materials.

  • Chemistry: Anion-exchange
  • Separation mode: Ion-exchange
  • Particle substrate: Polymethacrylate
  • pH range: 2 pH to 12 pH
  • Silanol activity: Low
  • Molecular weight range: 1000 to 500000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 1000 Å
  • Format: AP glass column
  • System: HPLC
  • USP classification: L23
  • Inner diameter: 8 mm
  • Length: 75 mm
  • Application: Protein
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.