Learn More
Waters Corp Waters Protein-Pak Hi Res HIC Column, 2.5 μm, 4.6 mm x 35 mm
Catalog No. 501528580 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Hi Res Hydrophobic Interaction Columns (HIC) are ideally suited for the characterization of recombinant proteins, monoclonal antibodies, and/or ADCs. They utilize non-porous particles with a low density of butyl ligands, providing outstanding resolution more quickly than many traditional porous HIC options.
  • Ideally suited for use in the characterization of recombinant proteins, monoclonal antibodies, and/or ADCs.
  • Uses non-porous particles containing a low density of butyl ligands, yielding outstanding resolution in less time compared to many traditional porous HIC offerings.
  • Quality control testing with defined protein standards helps ensure consistent batch-to-batch performance.
Catalog No. 50-152-8580 Supplier Waters Corp Supplier No. 186007582
These Hi Res Hydrophobic Interaction Columns (HIC) are ideally suited for the characterization of recombinant proteins, monoclonal antibodies, and/or ADCs. They utilize non-porous particles with a low density of butyl ligands, providing outstanding resolution more quickly than many traditional porous HIC options.
  • Ideally suited for use in the characterization of recombinant proteins, monoclonal antibodies, and/or ADCs.
  • Uses non-porous particles containing a low density of butyl ligands, yielding outstanding resolution in less time compared to many traditional porous HIC offerings.
  • Quality control testing with defined protein standards helps ensure consistent batch-to-batch performance.