Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein Phosphatase 1 Cata
Catalog No. 502904646
Encompass
Protein phosphatase1 (PP1) is a heterodimeric enzyme with serine/threonine phosphatase activity. It comprises a catalytic subunit and a targeting subunit or a specific protein inhibitor. The targeting subunit determines the enzyme localization e.g. to glycogen particles (subunit G) or to myofibrils (subunit M).
Catalog No. 50-290-4646 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P793725UG
Add to Cart
Add to Cart
Protein phosphatase1 (PP1) is a heterodimeric enzyme with serine/threonine phosphatase activity. It comprises a catalytic subunit and a targeting subunit or a specific protein inhibitor. The targeting subunit determines the enzyme localization e.g. to glycogen particles (subunit G) or to myofibrils (subunit M).