Learn More
Sino Biological Protein Pig ACVR2A 100
Catalog No. 5001074149
Encompass_Preferred
A DNA sequence encoding the Sus scrofa (Pig) ACVR2A (F1S0H6) (Met1-Pro135) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74149 Supplier Sino Biological Supplier No. 69111W08H100
A DNA sequence encoding the Sus scrofa (Pig) ACVR2A (F1S0H6) (Met1-Pro135) was expressed with a polyhistidine tag at the C-terminus