Learn More
Sino Biological Protein PLAUR ET-FREE 100UG
Catalog No. 5001074050
Encompass_Preferred
A DNA sequence encoding the human UPAR isoform 1 (Q03405-1) (Met 1-Arg 303) without the pro peptide was expressed with a carboxy-terminal polyhistidine tag
Catalog No. 50-010-74050 Supplier Sino Biological Supplier No. 10925H08HUE100
A DNA sequence encoding the human UPAR isoform 1 (Q03405-1) (Met 1-Arg 303) without the pro peptide was expressed with a carboxy-terminal polyhistidine tag