Learn More
BioVendor, LLC Pregnant Mare Serum Gonadotropin (PMSG), 5,000 IU
SDP
Supplier:  BioVendor, LLC RP1782725000
Encompass
TypeNative proteinDescriptionPMSG is a complex glycoprotein obtained from the serum of pregnant mares. This 43–63 kda protein is capable of supplementing and being substituted for the follicle stimulating gonadotrophin and interstitial cell-stimulating hormone of the anterior pituitary gland in both the male and female. Thus PMSG-Intervet stimulates development of the ovarian follicle in the female.SourceSerum of pregnant maresFormulationThe PMSG was lyophilized with no additives.ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationLyophilized PMSG although stable at room temperature for 3 weeks, should be stored between 2–8°C.Physical AppearanceSterile filtered white lyophilized (freeze-dried) powder.
Catalog No. 50-893-505