Learn More
Sino Biological Protein PODXL ET-FREE 100UG
Catalog No. 5001074033
Encompass_Preferred
A DNA sequence encoding the Human PODXL (O00592) (Ser23-Pro461) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-74033 Supplier Sino Biological Supplier No. 29798H08HUE100
A DNA sequence encoding the Human PODXL (O00592) (Ser23-Pro461) was expressed with a C-terminal polyhistidine tag