Learn More
Bioworld Protein pur-E.coli Lysis Buffer, 250 mL
Catalog No. 50253053
Encompass_Preferred
Protein Purification Buffer, E.coli Lysis Sample buffer for HIS-TAG protein purification from E.coli and insect cells. Technical Information: pH 8.0 Na2HPO4 (50mM)NaCl (300mM)Imidazole (10mM)
Catalog No. 50-253-053 Supplier Bioworld Supplier No. 220400511
Protein Purification Buffer, E.coli Lysis Sample buffer for HIS-TAG protein purification from E.coli and insect cells. Technical Information: pH 8.0 Na2HPO4 (50mM)NaCl (300mM)Imidazole (10mM)