Learn More
Macherey-Nagel Protein Quantification Assay , 50 reactions
Catalog No. NC1481596
Encompass_Preferred
Protein Quantification Assay (50) reagents for the protein quantification of 50 samples and 6 calibration curves with 7 calibration points each - Buffer PSB, Quantification Reagent, BSA
Catalog No. NC1481596 Supplier Macherey-Nagel Supplier No. 740967.50
Protein Quantification Assay (50) reagents for the protein quantification of 50 samples and 6 calibration curves with 7 calibration points each - Buffer PSB, Quantification Reagent, BSA