Learn More
Sino Biological Protein RABV-G
Catalog No. 5000131058
Encompass_Preferred
A DNA sequence encoding the Rabies virus (RABV) (strain CTN-1) Glycoprotein/G (FJ959397 1) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-001-31058 Supplier Sino Biological Supplier No. 41065V08H100
A DNA sequence encoding the Rabies virus (RABV) (strain CTN-1) Glycoprotein/G (FJ959397 1) was expressed with a C-terminal polyhistidine tag