Learn More
Sino Biological Protein Rat ICAM1 ET-FREE 1MG
Catalog No. 5001074101
Encompass_Preferred
A DNA sequence encoding the rat ICAM1 (Q00238-1) extracellular domain (Met 1-Thr 493) was expressed fused with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74101 Supplier Sino Biological Supplier No. 80022R08HUE1
A DNA sequence encoding the rat ICAM1 (Q00238-1) extracellular domain (Met 1-Thr 493) was expressed fused with a polyhistidine tag at the C-terminus