Learn More
Azzota Corp Heavy-Duty Portable Clinical Protein Refractometer. With Atc, 0-12 G/Dl, Refractive Index: Range: 1.330-1.360 Ri, Urine Specific Gravity: Range 1.00-1.050 Sg
Catalog No. 501179537
Encompass_Preferred
Heavy-Duty Portable Clinical Protein Refractometer. With ATC, 0-12 g/dl, Refractive Index: Range: 1.330-1.360 RI, Urine Specific Gravity: Range 1.00-1.050 SG
Catalog No. 50-117-9537 Supplier Azzota Corp Supplier No. RHCDL
Heavy-Duty Portable Clinical Protein Refractometer. With ATC, 0-12 g/dl, Refractive Index: Range: 1.330-1.360 RI, Urine Specific Gravity: Range 1.00-1.050 SG